Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Pocketed Hygiene Gown

Catalog No. 19910572 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-572 White 3X-Large
19-910-573 Blue Large
19-910-574 Blue X-Large
19-910-575 Blue 2X-Large
19-910-576 Blue 3X-Large
19-910-568 White Medium
19-910-569 White Large
19-910-570 White X-Large
19-910-571 White 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-910-572 Supplier Medicom™ Supplier No. 40.401/06
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketed Hygiene Gown with press-stud fasteners, available in two colors

  • Gown protects the operator, as well as sensitive products/processes that could be contaminated in a protected atmosphere
  • Protects user against radioactive particles, and infectious agents
  • Reduces the risk of soiling related to the user's activity
  • Pleasant and light, polypropylene fabric is breathable yet durable
  • Variety of colors allows for possibility of identification by color codes
  • Two pockets: one on the chest, and one on the lower thigh
  • Individually packaged for controlled hygiene, and distribution
  • Pockets located on the upper left, and lower right of the gown

Specifications

For Use With (Application) Category 1 Personal Protective Equipment
Size 3X-Large
Product Type Hygiene Gown
Packaging Type Individual, Bag
No. of Pockets 2
Gender Unisex
Construction Made in China
Material Polypropylene
Color White
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Resistance Radioactive Particles, Infectious Agents
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.