Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Polypropylene Coveralls

Catalog No. 19910624 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
Neck Style:
Collared
Hooded
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Neck Style Size
19-910-624 Blue Collared 3X-Large
19-910-621 Blue Collared Large
19-910-622 Blue Collared X-Large
19-910-623 Blue Collared 2X-Large
19-910-604 Blue Hooded Medium
19-910-605 Blue Hooded Large
19-910-606 Blue Hooded X-Large
19-910-607 Blue Hooded 2X-Large
19-910-601 White Hooded Small
19-910-602 White Hooded Large
19-910-603 White Hooded X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-910-624 Supplier Medicom™ Supplier No. 40.205BM/06
This item is not returnable. View return policy

Medicom™ Hopen™ Polypropylene Coveralls available in collared, or hooded styles

  • Hygiene Classic Polypropylene Coveralls protect the working environment from particles coming from the body of the operator
  • Protects the operator from the risks of soiling related to their purpose for use
  • Polypropylene fabric is light, and breathable
  • Elasticated at cuffs, waist, and ankles
  • Individually packaged for controlled hygiene and distribution
  • Category 1 Personal Protective Equipment

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Latex Free No
No. of Pockets None
For Use With (Application) Controlled Environments
Weight (Metric) 40 g/m2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.