Learn More
Structure Probe Inc HOPG SPI Supplies Brand Grade SPI-3 10x10x1 mm Thick, Package of 1 Piece
SDP
Supplier:  Structure Probe Inc 429HPAB
Encompass
Grade SPI-3. Exhibits a mosaic angle as small as 3.5° "+/- 1.5° and while it might not be acceptable for the most demanding of experiments, for student use its much more economical pricing makes it ideal. This grade is much less highly ordered and has a grain size not larger than the range of 30-40 nm. It is also excellent for student use. Because of the lower level of order, one can expect that the individual "strippings" will exhibit some population of "steps". Thickness may be ± 0.5 mm. Dimensions = 10 mm x 10 mm x 1 mm
Catalog No. NC1136001