Learn More
ABclonal Technology HOPX Rabbit PolymAb
SDP
Catalog No. 5000008205
Encompass_Preferred
The protein encoded by this gene is a homeodomain protein that lacks certain conserved residues required for DNA binding It was reported that choriocarcinoma cell lines and tissues failed to express this gene which suggested the possible involvement of this gene in malignant conversion of placental trophoblasts Studies in mice suggest that this protein may interact with serum response factor (SRF) and modulate SRF-dependent cardiac-specific gene expression and cardiac development Multiple alternatively spliced transcript variants have been identified for this gene
Catalog No. 50-000-08205 Supplier ABclonal Technology Supplier No. A15537PM20UL
The protein encoded by this gene is a homeodomain protein that lacks certain conserved residues required for DNA binding It was reported that choriocarcinoma cell lines and tissues failed to express this gene which suggested the possible involvement of this gene in malignant conversion of placental trophoblasts Studies in mice suggest that this protein may interact with serum response factor (SRF) and modulate SRF-dependent cardiac-specific gene expression and cardiac development Multiple alternatively spliced transcript variants have been identified for this gene