Learn More
AdipoGen Hordenin
SDP
Supplier:  AdipoGen CDXH0099G001
Encompass_Preferred
Chemical. CAS 539-15-1. Formula C10H15NO. MW 165.23. Synthetic. Hordenine is a phenethylamine type alkaloid found naturally in various plants such as barley and cacti. Hordenine may be used as an analytical reference material and in research on the activities of phenethylamine type alkaloids. Quorum-sensing inhibitor QSI.
Catalog No. 50-256-0740