Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Accessories: Sample Cups

Catalog No. 23600497 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Color:
Blue
Green
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-600-497 Yellow
23-600-494 Blue
23-600-495 Green
23-600-496 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-600-497 Supplier HORIBA Supplier No. 1220001768
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Pentra 400 Clinical Chemistry Analyzer

  • Available in multiple colors

Specifications

Type Sample Cup
For Use With (Equipment) ABX Pentra™ 400
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.