Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Controls

Catalog No. 23600413 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Type:
Immuno I Control L/H
Immuno II Control L/H
Quantity:
1 x 3 mL + 1 x 3 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
23-600-413 Immuno II Control L/H 1 x 3 mL + 1 x 3 mL
23-600-412 Immuno I Control L/H 1 x 3 mL + 1 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-600-413 Supplier HORIBA Supplier No. 1220001622
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE
For Use With (Equipment) ABX Pentra™ C400
Type Immuno II Control L/H
Intended Use Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin)
Level High, Low
Quantity 1 x 3 mL + 1 x 3 mL
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.