Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ISE Reagents for Pentra™ 400 Clinical Chemistry Analyzer

Catalog No. 23600483 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
100 mL
280 mL
Type:
Reference
Standard 1
Standard 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-600-483 100 mL Reference
23-600-481 280 mL Standard 1
23-600-482 100 mL Standard 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-600-483 Supplier HORIBA Supplier No. 1220001719
 Restricted Item
Add to Cart
Add to Cart

For use with Pentra 400 Clinical Chemistry Analyzer.

  • For use with Pentra 400 Clinical Chemistry Analyzer
  • Formulation: RTU
  • Container: Bottle

†Formulation (all items): RTU.

Specifications

Type Reference
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.