Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra 400 CO2 Substrate Reagent
GSA_VA

Catalog No. 23600701 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-600-701 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-600-701 Supplier HORIBA Supplier No. 1220001645
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with the Horiba ABX Pentra 400 Clinical Chemistry analyzer

  • Over 32 methods with applications for plasma, serum and urine
  • Less post-dilution due to greater linearity
  • Based on the latest research in Clinical Chemistry and complies with international recommendations (IFCC)
  • Standardized and in compliance with international recommendations
  • All specific proteins are standardized against CRM 470
  • Purified and concentrated antiserums
  • Long shelf-life (up to 5 years)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type CO2 RTU Substrate
Formulation UV Enzymatic
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.