Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Reagents for ABX Pentra C400
GSA_VA

Catalog No. 23601501 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Type:
CO2 Calibrator
CO2 Control
Reference Reagent
Form:
Ready-to-Use (RTU)
Quantity:
280 mL
3 x 3mL
4 x 3mL + 4 x 3mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Form Quantity
23-601-501 CO2 Control Ready-to-Use (RTU) 4 x 3mL + 4 x 3mL
23-601-500 CO2 Calibrator Ready-to-Use (RTU) 3 x 3mL
23-601-000 Reference Reagent Ready-to-Use (RTU) 280 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-601-501 Supplier HORIBA Supplier No. 5301101890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FDA cleared Liquid Chemistry

  • With extended linearity and increased stability
  • Standardized specific proteins
  • Purified and concentrated antiserums

Compliance

Complies with international recommendations (IFCC)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type CO2 Control
For Use With (Application) Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin)
For Use With (Equipment) ABX Pentra™ C400
Level High, Low
Quantity 4 x 3mL + 4 x 3mL
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.