Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Enzyme Assays for ABX Pentra™ 400 Clinical Chemistry Analyzers
GSA_VA

Catalog No. 23600418 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
100 Tests
125 Tests
250 Tests
Type:
ALP CP Reagent
ALT CP Reagent
AST CP Reagent
Amylase CP Reagent
CK NAC CP Reagent
GGT CP Reagent
Lipase CP Reagent
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-600-418 250 Tests ALT CP Reagent
23-600-417 125 Tests ALP CP Reagent
23-600-419 125 Tests Amylase CP Reagent
23-600-423 125 Tests CK NAC CP Reagent
23-600-421 250 Tests GGT CP Reagent
23-600-420 250 Tests AST CP Reagent
23-600-422 100 Tests Lipase CP Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 23-600-418 Supplier HORIBA Supplier No. 1220001627
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type ALT CP Reagent
Formulation IFCC
For Use With (Application) Quantitative determination of Alanine Aminotransferase in Serum or Plasma, in-vitro diagnostic use only
Includes 56mL R1 includes Tris, L-Alanine, Lactate Dehydrogenase, Sodium Azide, 14mL R2 includes 2-oxoglutarate, NADH, Sodium Azide
Quantity 250 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.