Learn More
Peak Scientific Instruments Ltd Horizen 24 Nitrogen Gas Generator Produces up to 24L/min of high purity nitrogen
SDP
Catalog No. NC2879248
Encompass_Preferred
The most energy-efficient nitrogen generator on the marketUltra-dry, non-methane hydrocarbon-free nitrogen at up to 99.9% purity and up to 116 psi55% less energy, saving power consumption*Reduction in heat output by over 50%, saving on air conditioning costs*2-year comprehensive manufacturer’s warranty**The smallest nitrogen generator in its class, fitting easily under any lab benchMinimized operational carbon footprint compared with cylinders and equivalent generator modelsState-of-the-art patented AirMax™ technology Easy to plan annual maintenance
Catalog No. NC2879248 Supplier Peak Scientific Instruments Ltd Supplier No. 3304532
Add to Cart
Add to Cart
Request a Quote
The most energy-efficient nitrogen generator on the marketUltra-dry, non-methane hydrocarbon-free nitrogen at up to 99.9% purity and up to 116 psi55% less energy, saving power consumption*Reduction in heat output by over 50%, saving on air conditioning costs*2-year comprehensive manufacturer’s warranty**The smallest nitrogen generator in its class, fitting easily under any lab benchMinimized operational carbon footprint compared with cylinders and equivalent generator modelsState-of-the-art patented AirMax™ technology Easy to plan annual maintenance