Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Horizon Fuel Cell H-Racer and H-Racer 2.0
SCI_ED

Catalog No. S98604
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98604 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98604 Supplier Horizon Fuel Cell Supplier No. FCJJ23

missing translation for 'validMainframeMsg'

Includes

Solar cell, solar refueling station, hydrogen fuel-cell car assembly parts, instruction manual, and Renewable Energy Education CD with science education and experiment manuals and assembly guide. H-Racer 2.0 also includes remote control with batteries.

Combination of hydrogen with oxygen generates a DC current to power an electric motor.

  • Unique miniature fuel-cell car and solar hydrogen refueling station
  • Refueling station creates hydrogen by electrolysis using water
  • Fill the hydrogen fuel tank and the car runs on its own fuel-cell system with the simple flip of a switch
  • H-Racer 2.0 features IR, remote-controlled steering
  • Grades 3+

Specifications

Grade 3+
For Use With (Application) For classroom
Includes Solar cell, solar refueling station, hydrogen fuel-cell car assembly parts, instruction manual, and renewable energy education CD with science education and experiment manuals and assembly guide. H-Racer 2 also includes remote control with batteries
Product Type H-racer2.0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.