Learn More
Chemglass Life Sciences Horizontal 15mL Tube Holder (12 Tubes) (Not Shown)
SDP
Catalog No. 5000083170
Encompass
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-000-83170 Supplier Chemglass Life Sciences Supplier No. CLS4891027
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.