Learn More
Grainger Horizontal Head For 12 X 1.5Ml
Catalog No. 502749848
Encompass_Preferred
The Benchmark Scientific Horizontal Head For 12 X 1.5Ml Tubes is a horizontal tube rotator designed for use with 1.5mL microcentrifuge tubes. It features a compact design with a low profile that allows tubes to be rotated at an angle between 5-80 degrees. The unit has adjustable speed up to 80rpm and accommodates up to 12 tubes at once. It requires a 12VDC power supply which is sold separately. This horizontal tube rotator provides gentle end-over-end agitation for applications such as dissolving, mixing, and resuspending samples in 1.5mL tubes.
Catalog No. 50-274-9848 Supplier Grainger Supplier No. 14N107
The Benchmark Scientific Horizontal Head For 12 X 1.5Ml Tubes is a horizontal tube rotator designed for use with 1.5mL microcentrifuge tubes. It features a compact design with a low profile that allows tubes to be rotated at an angle between 5-80 degrees. The unit has adjustable speed up to 80rpm and accommodates up to 12 tubes at once. It requires a 12VDC power supply which is sold separately. This horizontal tube rotator provides gentle end-over-end agitation for applications such as dissolving, mixing, and resuspending samples in 1.5mL tubes.