Learn More
Air Science Usa Horizontal Laminar Flow Cabinet Clean Bench, 48" / 1200mm Nominal Width, 115V 60Hz
SDP
Catalog No. NC1837392
Encompass_Preferred
Purair Lf Series Horizontal Laminar Flow Cabinet / Clean Bench (hlf-48), 48" / 1200mm Nominal Width, 115V 60Hz (other voltages available). Purair Horizontal Laminar Flow Hoods are a series of high efficiency products designed to protect equipment and other contents of the work zone from particulates, for applications sensitive to such contamination. It is ideally suited for use with non-hazardous contaminants and when flexible access to the equipment in the work zone is desired. At the heart of the Purair Laminar Flow Hood product line is the Multiplex ULPA Filtration Technology that creates a clean work environment over a wide range of applications
Catalog No. NC1837392 Supplier Air Science Usa Supplier No. HLF48
Purair Lf Series Horizontal Laminar Flow Cabinet / Clean Bench (hlf-48), 48" / 1200mm Nominal Width, 115V 60Hz (other voltages available). Purair Horizontal Laminar Flow Hoods are a series of high efficiency products designed to protect equipment and other contents of the work zone from particulates, for applications sensitive to such contamination. It is ideally suited for use with non-hazardous contaminants and when flexible access to the equipment in the work zone is desired. At the heart of the Purair Laminar Flow Hood product line is the Multiplex ULPA Filtration Technology that creates a clean work environment over a wide range of applications