Learn More
Electron Microscopy Sciences Secador™ Horizontal Shelves for Desiccator Cabinets
SDP
Catalog No. 501899534
Encompass_Preferred
To convert your Model 4.0 Secador Autodesiccator Cabinets or Secador Desiccator Cabinets from the Horizontal configuration to the Vertical, or from Vertical configuration to Horizontal, all you need is a new set of shelves. 2 shelves.
Catalog No. 50-189-9534 Supplier Electron Microscopy Sciences Supplier No. 7122910
To convert your Model 4.0 Secador Autodesiccator Cabinets or Secador Desiccator Cabinets from the Horizontal configuration to the Vertical, or from Vertical configuration to Horizontal, all you need is a new set of shelves. 2 shelves.