Learn More
Azzota Corp Stage Micrometers Slide (Stage Graticules), Horizontal, Total Length Of Micrometer Scale: 1 mm, 0.01 mm Per Division, Accuracy: <=3%, Ideal For Microscope Calibration
Supplier:  Azzota Corp TSM1
Encompass_Preferred
Stage Micrometers Slide (Stage Graticules), Horizontal, Total length of micrometer scale: 1 mm, 0.01 mm per division, Accuracy: <=3%, Ideal for microscope calibration
Catalog No. 50-753-2911