Learn More
PicoStudio Horizontal Turbine
SCI_ED
Catalog No. S4131909
Change view
Click to view available options
Description:
Level 1
Level 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
S4131909 Level 3
S4131907 Level 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Small Parts - Not recommended for children under 3 years old.
Catalog No. S4131909 Supplier PicoStudio Supplier No. PS30013
Add to Cart
Add to Cart
Small Parts - Not recommended for children under 3 years old.
The Horizontal Turbine teaches how wind turbines are designed and built, and how they generate energy.
  • Explore the power of wind energy with our Horizontal Wind Turbine!
  • Learn how wind turbines are designed and built, and how they generate energy.
Specifications
Product Type Kit, STEM
Description Level 3
Includes Horizontal Level 2 Parts, 1 AC Generator Pack, 1 Rectification Lab, 1 Long Tower, 1 Weighted Base Insert, 1 Circuit 101 Lab (3 Resistors 10, 30, 100 Ohm, 2 AA Battery Holder, 1 Light Bulb 1.5V, 8 Male End Wires, 1 Light Bulb Stand, 1 Alligator Clips Set (Red and Black), 1 Multimeter), 1 Solar Panel 7.2V 200mA
Height (English) 18.5 in.
Height (Metric) 470 mm
Length (English) 15.5 in.
Length (Metric) 393.7 mm