Learn More
LaMotte™ Horizontal Water Sampler
SDPSCI_ED
Catalog No. S37728 Shop All LaMotte Company Products
Change view
Click to view available options
For Use With (Application):
Allows water to be sampled at specific depths
Catalog No. For Use With (Application)
S37728 Allows water to be sampled at specific depths
1 options
Catalog No. S37728 Supplier LaMotte™ Supplier No. 1087
Add to Cart
Add to Cart
Collect water samples quickly and easily!
  • Rugged stainless steel components
  • Suitable for sampling fresh, brackish or salt water
  • For educational use
  • High impact grad plastic tube holds approximately 1200 mL of sample and allows for visual inspection of sample prior to dispensing
Specifications
Product Type Horizontal Water Sampler
Includes 20m calibrated line
Material Stainless Steel
For Use With (Application) Allows water to be sampled at specific depths