Learn More
AdipoGen Hormaomycin
SDP
Supplier:  AdipoGen BVT0107C500
Encompass_Preferred
Chemical. CAS 121548-21-8 and 92092-69-8. Formula C55H69N10O14Cl. MW 1129.7. Isolated from Streptomyces griseoflavus. Antibiotic. Peptide lactone. Antibacterial agent selective. Potent narrow-spectrum antibiotic against coryneform actinomycetes MIC value 88 pM. Quorum sensing modulator. Shows hormonal activity. Inducer of morphological differentiation in actinomycetes. Stimulator of antibiotic production. Inducer of aerial mycelium formation in various actinomycetes. Antimalarial in vitro. Rare type of natural product, regulating bacterial behavior and independently function in a defensive antibiotic role.
Catalog No. 50-256-2095