Learn More
Cusabio Technology LLC Plant hormone abscisic acid,ABA ELISA Kit
Supplier:  Cusabio Technology LLC CSBE09159PL
Encompass_Preferred
This Plant ABA ELISA Kit was designed for the quantitative measurement of Plant ABA protein in plant tissues. It is a Competitive ELISA kit, its detection range is 0.156 渭g/mL-10 渭g/mL and the sensitivity is 0.04 渭g/mL. One kit contains 96 wells.
Catalog No. NC2952857