Learn More
OriGene Hormone sensitive lipase (LIPE) CytoSection, 25 Slides
SDP
Catalog No. NC3734795
Encompass_Preferred
Transient overexpression of LIPE in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3734795 Supplier OriGene Supplier No. TS409729P5
Transient overexpression of LIPE in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack