Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hormones: b-Estradiol

Catalog No. ICN19456505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 mg
5000 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19456505 5000 mg
ICN19456501 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19456505 Supplier MP Biomedicals™ Supplier No. 0219456505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For research use only. Not intended for therapeutic use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Crystalline powder
Product Type Beta-Estradiol
Sterility Sterile
Quantity 5000 mg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.