Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hormones: Hydrocortisone

Catalog No. ICN19456905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5mg
1 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19456905 5mg
ICN19456910 10 g
ICN19456925 25 g
ICN19456901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19456905 Supplier MP Biomedicals™ Supplier No. 0219456905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For research use only. Not intended for therapeutic use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Hydrocortisone
Sterility Sterile
Quantity 5mg
Source Soy beans
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.