Learn More
Cayman Chemical HormoneSensitive Lipase Polyc
SDP
Catalog No. 502886557
Encompass_Preferred
Immunogen: synthetic peptide from the C-terminal region of human HSL receptor • Species Reactivity: Human, mouse, rat HSL receptor • Host: rabbit • Application: WB
Catalog No. 50-288-6557 Supplier Cayman Chemical Supplier No. 100063711 EA
Immunogen: synthetic peptide from the C-terminal region of human HSL receptor • Species Reactivity: Human, mouse, rat HSL receptor • Host: rabbit • Application: WB