Learn More
BRANSON ULTRASONICS CORPORATION (3P) HORN CUP 2INCH
Catalog No. NC3721558
Encompass_Preferred
Branson Ultrasonics Sonifier Cup Horns, For Use With: Ideal for liposome preparation, viruses and radioactive materials, Sonifier S-450 cell disruptor, Size: 2 in., Diameter: 2 in., 5.08cm, Titanium
Catalog No. NC3721558 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 101147047
Branson Ultrasonics Sonifier Cup Horns, For Use With: Ideal for liposome preparation, viruses and radioactive materials, Sonifier S-450 cell disruptor, Size: 2 in., Diameter: 2 in., 5.08cm, Titanium