Learn More
BRANSON ULTRASONICS CORPORATION (3P) HORN DISRUPTOR F/HOMOGENZ .5IN
Catalog No. NC3721555
Encompass_Preferred
Branson Ultrasonics Sonifier Disruptor Horns, 1/2 in. dia., tapped configuration, step horn with threaded body, Amplitude Range: 21 to 145um, Diameter Tip: 0.5 in., 1.3cm, Intensity: High, Tip Style: 1/2 in. (13mm), Capacity: 10 to 250mL
Catalog No. NC3721555 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 101147037
Branson Ultrasonics Sonifier Disruptor Horns, 1/2 in. dia., tapped configuration, step horn with threaded body, Amplitude Range: 21 to 145um, Diameter Tip: 0.5 in., 1.3cm, Intensity: High, Tip Style: 1/2 in. (13mm), Capacity: 10 to 250mL