Learn More
Grainger Horn Red Surface Mounting
Catalog No. 502766665
Encompass_Preferred
The Edwards Signaling 24V DC Horn is a weatherproof audible signaling device designed for fire alarm, security, and process control applications. It produces a loud, penetrating sound and features a rugged ABS plastic housing, stainless steel hardware, and weatherproof construction for indoor or outdoor use. This horn operates on 24V DC and draws 0.18A. It has an adjustable volume between 80 to 103 dB at 10 feet and includes a screw terminal connection block. The compact horn measures 5.5" H x 5" W x 3.25" D and weighs 2 lbs. UL listed for fire protective service.
Catalog No. 50-276-6665 Supplier Grainger Supplier No. 16X295
The Edwards Signaling 24V DC Horn is a weatherproof audible signaling device designed for fire alarm, security, and process control applications. It produces a loud, penetrating sound and features a rugged ABS plastic housing, stainless steel hardware, and weatherproof construction for indoor or outdoor use. This horn operates on 24V DC and draws 0.18A. It has an adjustable volume between 80 to 103 dB at 10 feet and includes a screw terminal connection block. The compact horn measures 5.5" H x 5" W x 3.25" D and weighs 2 lbs. UL listed for fire protective service.