Learn More
Grainger Edwards Signaling Horn And Strobe, 24 V AC/DC, Xenon, 92 dB, 1 ea
Catalog No. 502792379
Encompass_Preferred
This horn strobe is a dual-function signaling device designed for indoor applications, combining an audible horn with a high-intensity xenon strobe light to provide effective audio and visual alerts. It operates on 24 V AC or 24 V DC power and is engineered for surface wall mounting, specifically designed to fit a standard single gang box.
  • Produces a loud horn tone with 92 dB sound output at 1 m (82 dB at 10 ft.)
  • Features a high-intensity xenon strobe with a 50 candela light output
  • Operates at a flash rate of 60 flashes per minute
  • Constructed with a durable thermoplastic housing in a neutral white finish
  • Supports continuous sound output for reliable signaling
  • Rated for indoor use in assistance call systems
  • UL 1638 listed for safety and compliance
Catalog No. 50-279-2379 Supplier Grainger Supplier No. 16G823
This horn strobe is a dual-function signaling device designed for indoor applications, combining an audible horn with a high-intensity xenon strobe light to provide effective audio and visual alerts. It operates on 24 V AC or 24 V DC power and is engineered for surface wall mounting, specifically designed to fit a standard single gang box.
  • Produces a loud horn tone with 92 dB sound output at 1 m (82 dB at 10 ft.)
  • Features a high-intensity xenon strobe with a 50 candela light output
  • Operates at a flash rate of 60 flashes per minute
  • Constructed with a durable thermoplastic housing in a neutral white finish
  • Supports continuous sound output for reliable signaling
  • Rated for indoor use in assistance call systems
  • UL 1638 listed for safety and compliance