Learn More
Grainger Edwards Signaling Horn and Strobe, 120V AC, Surface Mount, Clear, 1 ea
Catalog No. 502791780
Encompass_Preferred
The 860 Series Hybrid horn strobe is designed for indoor signaling applications requiring a high-decibel audible alert combined with a visual xenon strobe. This surface-mount unit features a durable gray flame-resistant thermoplastic housing and produces a continuous horn sound at 90 dB measured at 10 ft. (100 dB at 1 m) with 60 flashes per minute.
  • Operating voltage of 120V AC
  • Low current draw of 148 mA
  • Clear strobe lens with 150 CP light intensity
  • Designed for surface mounting with included back box
  • Operating temperature range of +32°F to +120°F
  • UL Listed
Catalog No. 50-279-1780 Supplier Grainger Supplier No. 16G545
The 860 Series Hybrid horn strobe is designed for indoor signaling applications requiring a high-decibel audible alert combined with a visual xenon strobe. This surface-mount unit features a durable gray flame-resistant thermoplastic housing and produces a continuous horn sound at 90 dB measured at 10 ft. (100 dB at 1 m) with 60 flashes per minute.
  • Operating voltage of 120V AC
  • Low current draw of 148 mA
  • Clear strobe lens with 150 CP light intensity
  • Designed for surface mounting with included back box
  • Operating temperature range of +32°F to +120°F
  • UL Listed