Learn More
Grainger Edwards Signaling 860 Industrial Horn Strobe, 24 VAC/VDC, Xenon, Surface Mount, 1 ea
Catalog No. 502791986
Encompass_Preferred
This industrial electronic horn and xenon strobe is a combination signal designed for environments where a distinct visual or audible alarm is required. Built for durability, it features a corrosion-resistant, weatherproof surface box suitable for outdoor applications. The unit provides high-decibel sound output with a bright strobe and allows for independent operation of the horn and light.
  • Designed for outdoor use with a temperature range of -31°F to 150°F
  • Features 100/90 dB sound level and 150 cd light output
  • Includes corrosion-resistant, weatherproof surface box
  • Allows independent operation of strobe and horn
  • Operates on 24 VAC/VDC
  • Terminals provided for easy wiring
  • UL 1638 and UL 464 listed and RoHS compliant
Catalog No. 50-279-1986 Supplier Grainger Supplier No. 16G549
This industrial electronic horn and xenon strobe is a combination signal designed for environments where a distinct visual or audible alarm is required. Built for durability, it features a corrosion-resistant, weatherproof surface box suitable for outdoor applications. The unit provides high-decibel sound output with a bright strobe and allows for independent operation of the horn and light.
  • Designed for outdoor use with a temperature range of -31°F to 150°F
  • Features 100/90 dB sound level and 150 cd light output
  • Includes corrosion-resistant, weatherproof surface box
  • Allows independent operation of strobe and horn
  • Operates on 24 VAC/VDC
  • Terminals provided for easy wiring
  • UL 1638 and UL 464 listed and RoHS compliant