Learn More
Grainger Edwards Signaling Horn and Strobe, 120V AC, Red Lens, Surface Mount, 1 ea
Catalog No. 502792237
Encompass_Preferred
These combination horn and strobe beacons provide high-decibel audible signaling and high-visibility visual alerts for indoor or outdoor applications. Designed for industrial use, this unit features independent operation of the horn and xenon strobe and is constructed with a durable gray thermoplastic housing.
  • Operating voltage of 120V AC
  • Sound output of 100 dB at 1 m (90 dB at 10 ft.)
  • Light output of 21 cd (effective)
  • Flash rate of 60 fpm
  • Surface mount design
  • Diode polarized for use in electrically supervised circuits
  • Operating temperature range of -31°F to 150°F
  • UL listed
Catalog No. 50-279-2237 Supplier Grainger Supplier No. 16G542
These combination horn and strobe beacons provide high-decibel audible signaling and high-visibility visual alerts for indoor or outdoor applications. Designed for industrial use, this unit features independent operation of the horn and xenon strobe and is constructed with a durable gray thermoplastic housing.
  • Operating voltage of 120V AC
  • Sound output of 100 dB at 1 m (90 dB at 10 ft.)
  • Light output of 21 cd (effective)
  • Flash rate of 60 fpm
  • Surface mount design
  • Diode polarized for use in electrically supervised circuits
  • Operating temperature range of -31°F to 150°F
  • UL listed