Learn More
Ace Glass, Inc. Horn, ultrasonic, 1/2in, threaded tip
SDP
Catalog No. 50980544
Encompass
1/2 ultrasonic titanium horn (probe) with a threaded end, that focuses the ultrasonic energy into the liquid. Fabricated from high grade titanium, these horns are autoclavable and have an O-ring groove at the nodal point that allows a tight fit in a #36 Ace-Thred without affecting sonic output. The threaded end accepts replaceable tips, microtips, or extenders. Supplied with a 1/2-20 stud for connection to the converter on a power supply.
Catalog No. 50-980-544 Supplier Ace Glass, Inc. Supplier No. 981425
1/2 ultrasonic titanium horn (probe) with a threaded end, that focuses the ultrasonic energy into the liquid. Fabricated from high grade titanium, these horns are autoclavable and have an O-ring groove at the nodal point that allows a tight fit in a #36 Ace-Thred without affecting sonic output. The threaded end accepts replaceable tips, microtips, or extenders. Supplied with a 1/2-20 stud for connection to the converter on a power supply.