Learn More
Innovative Research Inc Horse Albumin Pur Lyo 100mg
Supplier:  Innovative Research Inc IHSALBLY100MG
Encompass_Preferred
Horse Albumin Purified Lyophilized 100mg - Horse Albumin Purified Lyophilized from Innovative Research has been purified from horse plasma by a propritetary process that combines multiple conventional chromatographic methods. This is a Lyophilized powder buffered in Salt free and a purity of 95% as determined by SDS-PAGE. More Details Species Horse Target Albumin Purification Method Chromatographically purified Purity 95% as determined by SDS-PAGE Analysis Source Horse plasma Storage Conditions 4 C Additional Information Add deionized water or buffer to original or desired volume, aliquot and freeze unused portion. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including blocking, cell signaling, drug studies, protein-protein interactions, protein stabilization, receptor binding, tissue culture.
Catalog No. 50-274-4366