Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Horse anti-Mouse IgG (H+L) Secondary Antibody

Catalog No. PI31181 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31181 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31181 Supplier Invitrogen™ Supplier No. 31181
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Horse Polyclonal Secondary Antibody

Product # 31181 has been successfully used in Western blot, IF, ICC, IHC, ELISA and ISH applications. Product # 31181 reacts with heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins. Based on solid-phase enzyme immunoassay, this antibody cross-reacts with rat IgG and mouse IgM (25%), hamster IgG (12%), guinea pig IgG and rat IgM (6%), bovine IgM, dog IgM, goat IgM, human IgM, Rhesus monkey IgG, sheep IgG, sheep IgM, swine IgG (1%). It has minimum cross-reactivity with immunoglobulins from other species. The activity measured by a solid phase binding assay is 94%.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG (H+L)
Applications ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 20mg/mL sucrose and 0.08% sodium azide; pH 7.8
Host Species Equine
Purification Method Affinity chromatography
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.