Learn More
Horse anti-Rabbit IgG ImmPRESS(TM) Secondary Antibody [HRP Polymer], Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG ImmPRESS(TM) |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In Situ Hybridization (ISH) |
| Classification | Polyclonal |
| Conjugate | HRP Polymer |
| Host Species | Equine |
| Immunogen | Rabbit IgG |
| Purification Method | Affinity purified |
| Quantity | 15 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.