Learn More
Vector Laboratories Horse Anti-Goat IgG Antibody HRP 1 mg
SDP
Supplier:  Vector Laboratories PI95001
Encompass
This high-affinity horse anti-goat IgG secondary antibody is purified by affinity chromatography and adsorbed to remove any cross-reactivities that are likely to interfere with specific labeling. This antibody is then cross-linked with high specific activity horseradish peroxidase (HRP) using a proprietary conjugation procedure that preserves maximum enzyme activity and antibody specificity. This antibody is subjected to rigorous quality control assays and can be used for tissue staining, ELISAs, and blots. In addition to recognizing goat IgG, this antibody detects sheep and bovine IgG primary antibodies.
Catalog No. NC9586286