Learn More
Biomatik Corporation Horse Atrial Natriuretic Peptide (ANP) ELISA Kit
Supplier:  Biomatik Corporation EKU08674
Encompass_Preferred
Horse Atrial Natriuretic Peptide (ANP) ELISA Kit for detecting ANP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 24.69-2000pg/ml. Sensitivity: 9.17pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P27104. Gene ID: 100034201. Target Synonyms: ANH; ANF; CDD; Atrial Natriuretic Factor; Atrial Natriuretic Hormone; Atriopeptin; Cardionatrine; Cardiodilatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8636