Learn More
American Research Products Inc Horse Beta Endorphin b EP ELI
Catalog No. 5000326480
Encompass
Horse Beta-Endorphin b-EP ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26480 Supplier American Research Products Inc Supplier No. CSBE06831EQ
Horse Beta-Endorphin b-EP ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma Method Sandwich