Learn More
Biomatik Corporation Horse Beta-Endorphin (bEP) ELISA Kit
Supplier:  Biomatik Corporation EKU08700
Encompass_Preferred
Horse Beta-Endorphin (bEP) ELISA Kit for detecting bEP in serum, plasma, cerebrospinal fluid, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.03pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A3Q2IBW0. Target Synonyms: b-EP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4483