Learn More
Hemostat Laboratories HORSE BLOOD DEFIB 500ML
SDP
Catalog No. 50863761
Encompass_Preferred
Defibrinated Blood - Horse 500Ml. Aseptically Collected From Healthy Animals Of U.S. Origin. Mechanically Defibrinated Without Anticoagulants Or Other Additives. Proprietary Sceening Process Insures Low Fragility And Consistent Color.
Catalog No. 50-863-761 Supplier Hemostat Laboratories Supplier No. DHB500
Defibrinated Blood - Horse 500Ml. Aseptically Collected From Healthy Animals Of U.S. Origin. Mechanically Defibrinated Without Anticoagulants Or Other Additives. Proprietary Sceening Process Insures Low Fragility And Consistent Color.