Learn More
American Research Products Inc Horse Erythropoietin EPO ELI
Catalog No. 5000328086
Encompass
Horse Erythropoietin (EPO) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28086 Supplier American Research Products Inc Supplier No. CSBEL007743HO
Horse Erythropoietin (EPO) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich