Learn More
Biomatik Corporation Horse Hemoglobin (HB) ELISA Kit
Supplier:  Biomatik Corporation EKU04713
Encompass_Preferred
Horse Hemoglobin (HB) ELISA Kit for detecting HB in serum, plasma, erythrocyte lysates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 9.38-600ng/ml. Sensitivity: 3.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01958; P02062; P55102; P06714. Gene ID: 100034076; 100036557. Target Synonyms: Hgb; Haemoglobin; Heterotetramer (αβ) 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8935