Learn More
American Research Products Inc Horse hypoXia inducible factor
Catalog No. 5000326898
Encompass
Horse hypoXia inducible factor 1 alpha subunit (basic heliX-loop-heliX transcription factor) (HIF1A) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26898 Supplier American Research Products Inc Supplier No. CSBEL010351HO
Horse hypoXia inducible factor 1 alpha subunit (basic heliX-loop-heliX transcription factor) (HIF1A) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich