Learn More
Biomatik Corporation Horse Insulin (INS) ELISA Kit
Supplier:  Biomatik Corporation EKU11568
Encompass_Preferred
Horse Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 123.5-10,000pg/ml. Sensitivity: 53.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01310. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6975