Learn More
American Research Products Inc Horse insulin like growth fact
Catalog No. 5000327234
Encompass
Horse insulin-like growth factor 1 (IGF1) ELISA kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27234 Supplier American Research Products Inc Supplier No. CSBEL011086HO
Horse insulin-like growth factor 1 (IGF1) ELISA kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich