Learn More
American Research Products Inc Horse Interleukin 6 IL 6 ELI
Supplier:  American Research Products Inc CSBE16634HS
Encompass
Horse Interleukin 6 (IL-6) ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26809