Learn More
American Research Products Inc Horse Interleukin 8 IL 8 ELIS
Catalog No. 5000330101
Encompass
Horse Interleukin 8 IL-8 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30101 Supplier American Research Products Inc Supplier No. CSBE14021HS
Add to Cart
Add to Cart
Horse Interleukin 8 IL-8 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich