Learn More
Cusabio Technology LLC Horse LeptinLEP ELISA kit
Catalog No. 5000049593
Encompass_Preferred
The Horse Leptin (LEP) ELISA Kit is designed to detect and quantify LEP concentrations in equine serum plasma and tissue homogenates with high sensitivity and precision This kit employs a competitive ELISA format enabling reliable quantitative measurement within a dynamic range of 3 12 pg/mL to 200 pg/mL and achieves a lower detection limit of 0 78 pg/mL The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours with absorbance readings taken at 450 nm Suitable for both research and diagnostic applications it provides a standardized user-friendly method to assess leptin levels in horses ensuring consistent reproducibility across experimental or clinical settings
Catalog No. 50-000-49593 Supplier Cusabio Technology LLC Supplier No. CSBEL012870HO
The Horse Leptin (LEP) ELISA Kit is designed to detect and quantify LEP concentrations in equine serum plasma and tissue homogenates with high sensitivity and precision This kit employs a competitive ELISA format enabling reliable quantitative measurement within a dynamic range of 3 12 pg/mL to 200 pg/mL and achieves a lower detection limit of 0 78 pg/mL The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours with absorbance readings taken at 450 nm Suitable for both research and diagnostic applications it provides a standardized user-friendly method to assess leptin levels in horses ensuring consistent reproducibility across experimental or clinical settings